Silicon standard solution, 10000 mg/L Si in water with hydrofluoric acid (max. 1%) (from (NH₄)₂SiF₆), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
38723.AD 38723.AE 38723.AP
38723.ADEA 131 EUR
38723.AD 38723.AE 38723.AP
Silicon standard solution, 10000 mg/L Si in water with hydrofluoric acid (max. 1%) (from (NH₄)₂SiF₆), standard for ICP, Specpure®
Standaarden Kationische standaarden Silicium standaarden
Molecular Formula: (NH₄)₂SiF₆ in H₂O/tr. F-
Physical Form: Liquid
MDL No.: MFCD00085311
Certificaten: Traceable to NIST