Tin standard solution, 1000 mg/L Sn in 20% hydrochloric acid (from Sn), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13863.AD
13863.AE
13863.AP
13863.ADEA
70.3
EUR
13863.AD
13863.AE
13863.AP
Tin standard solution, 1000 mg/L Sn in 20% hydrochloric acid (from Sn), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Tin standaarden
Molecular Formula: Sn in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00133862
Certificaten: Traceable to NIST