Tin standard solution, 1000 mg/L Sn in 20% hydrochloric acid (from Sn), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88112.AE
88112.AP
88112.AEEA
48
EUR
88112.AE
88112.AP
Tin standard solution, 1000 mg/L Sn in 20% hydrochloric acid (from Sn), standard for AAS, Specpure®
Standaarden
Kationische standaarden
Tin standaarden
Molecular Formula: Sn in 20% HCl
Physical Form: Liquid
Odour: Characteristic
UN No.: 1789
MDL No.: MFCD00133862
Certificaten: Traceable to NIST