Yttrium standard solution, 1000 mg/L Y in 5% nitric acid (from Y₂O₃), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88117.AP 88117.AE
88117.APEA 101 EUR
88117.AP 88117.AE
Yttrium standard solution, 1000 mg/L Y in 5% nitric acid (from Y₂O₃), standard for AAS, Specpure®
Standaarden Kationische standaarden Yttrium standaarden
Molecular Formula: Y₂O₃ in 5% HNO₃
Physical Form: Liquid
Odour: Characteristic
UN No.: 3264
MDL No.: MFCD00011468
Certificaten: Traceable to NIST