Laboratory glass cleaner, deconex® 15 NF
Leverancier: Borer Chemie
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
514500.00-F10W 514500.00-K05W 514500.00-K10W
DECO514500.00-F10WEA 24.1 EUR
DECO514500.00-F10W DECO514500.00-K05W DECO514500.00-K10W
Laboratory glass cleaner, deconex® 15 NF
Reinigingsmiddelen Detergenten en oppervlaktereinigers
Liquid, alkaline cleaning concentrate for manual cleaning of laboratory glassware and reusable laboratory materials. Is especially suitable for the pre-cleaning before automated cleaning - also without intermediate rinsing.
  • Rinsable without residues and guarantees therefore consistent and error-free results
  • With a broad cleaning spectrum and perfect results even at low concentrations
  • Working solution suitable for multiple use
  • Facilitates the complete detachment of stubborn deposits in automated cleaning
  • Residues carried over from pre-cleaning do not cause foaming in laboratory washers
  • Phosphate-free, suitable for phosphate-critical applications
  • EDTA-, NTA- and chlorine-free, environmentally friendly
Suitable for soaking baths, pipette washers and ultrasonic devices.