Laboratory glass cleaner, deconex® 22 HPF-x
Leverancier: Borer Chemie
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
516900.00-KK5W 516900.00-K10W 516900.00-K25W
DECO516900.00-KK5WEA 86 EUR
DECO516900.00-KK5W DECO516900.00-K10W DECO516900.00-K25W
Laboratory glass cleaner, deconex® 22 HPF-x
Reinigingsmiddelen Detergenten en oppervlaktereinigers
Liquid, highly alkaline cleaning concentrate for automated cleaning of laboratory glassware und reusable laboratory material.
  • Rinsable without residues and guarantees therefore consistent and error-free results
  • With a broad cleaning spectrum and excellent results even at low concentrations
  • High cleaning performance due to excellent dispersing capabilities, effective against stubborn deposits
  • Well suited for an effective cleaning of animal cages
  • Phosphate- and surfactant-free, no release of phosphates and surfactants into waste water
  • EDTA-, NTA- and chlorine-free, environmentally friendly
  • Rinsing with an acidic deconex® cleaner is recommended as the completion of the cleaning process