Gold standard solution, 1000 mg/L Au in 2 M hydrochloric acid (from HAuCl?) Certipur® standard for AAS, Supelco®
Leverancier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.70216.0100
1.70216.0500
1.70216.0100EA
137
EUR
1.70216.0100
1.70216.0500
Gold standard solution, 1000 mg/L Au in 2 M hydrochloric acid (from HAuCl?) Certipur® standard for AAS, Supelco®
Standaarden
Kationische standaarden
Goud standaarden
Certificaten: Traceable to SRM from NIST