Phenol : chloroform : iso-amyl alcohol (125:24:1) (v/v/v), BioUltra for molecular biology, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar


M-Clarity™ quality level = MQ100

77619-100ML 77619-500ML
SUPL77619-100MLEA 108 EUR
SUPL77619-100ML SUPL77619-500ML
Phenol : chloroform : iso-amyl alcohol (125:24:1) (v/v/v), BioUltra for molecular biology, Supelco®
Phenol : Chloroform : iso-Amyl alcohol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00133763
CAS nummer: 136112-00-0
UN: 2810
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73fce16b08856220951f4da3d5fee1e6d01511a2&vid=2a16f5391f26207f0b4265b94a167cc1c614f420&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR