Trioctylamine ≥97.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
T0502-25ML
T0502-500ML
T0502-100ML
TCIAT0502-25MLEA
19.8
EUR
TCIAT0502-25ML
TCIAT0502-500ML
TCIAT0502-100ML
Trioctylamine ≥97.0% (by GC, titration analysis)
Trioctylamine
Formule:
C₂₄H₅₁N Kookpunt: 367°C Smeltpunt: –39 °C Dichtheid: 0,812 g/cm³ (20 °C) Vlampunt: 184°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00009560 CAS nummer: 1116-76-3 EINECS: 214-242-1 ADR: 9,III |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...