2-Chloro-4-nitrotoluene ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
C0690-500G C0690-25G
TCIAC0690-500GEA 208 EUR
TCIAC0690-500G TCIAC0690-25G
2-Chloro-4-nitrotoluene ≥99.0% (by GC)
2-Chloro-4-nitrotoluene
Formule: C₇H₆ClNO₂
MW: 171.58 g/mol
Kookpunt: 260°C
Smeltpunt: 68
MDL Nummer: MFCD00007210
CAS nummer: 121-86-8
EINECS: 204-501-7
UN: 3457
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2aba2df7400a85e348ca0dc8d1dbfe51249e126&vid=4f9363ac02e64d144885e1cb2b37e3d60c007c58&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity (GC) min. 99.0 %
Melting point 62.0 to 65.0 deg-C