Potassium hydrogen carbonate, Reagent Grade
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Potassium bicarbonate
0889-1KGEA 175 EUR
0889-1KG 0889-500G
Potassium hydrogen carbonate, Reagent Grade
Potassium hydrogen carbonate
Formule: KHCO₃
MW: 100.12 g/mol
Smeltpunt: 292
Dichtheid: 2.17 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011402
CAS nummer: 298-14-6
EINECS: 206-059-0
Merck Index: 13,07691