Triethyl acetylcitrate ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
O-Acetylcitric acid triethyl ester, Triethyl O-Acetylcitrate, Acetyltriethylcitrate, Triethyl 2-acetoxypropane-1,2,3-tricarboxylate
A0086-25G
A0086-500G
TCIAA0086-25GEA
17.6
EUR
TCIAA0086-25G
TCIAA0086-500G
Triethyl acetylcitrate ≥97.0% (by GC)
Triethyl acetylcitrate
Formule:
C₁₄H₂₂O₈ MW: 318.32 g/mol Kookpunt: 229°C Vlampunt: 188°C |
MDL Nummer:
MFCD00049378 CAS nummer: 77-89-4 EINECS: 201-066-5 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...