Cesium standard solution, 10000 mg/L Cs in 5% nitric acid (from Cs₂CO₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14425.AD 14425.AE 14425.AP
14425.ADEA 100 EUR
14425.AD 14425.AE 14425.AP
Cesium standard solution, 10000 mg/L Cs in 5% nitric acid (from Cs₂CO₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Cesium standaarden
Molecular Formula: Cs₂CO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134037
Certificaten: Traceable to NIST
Order Now
Specificaties
Environmentally Preferable
MDL No. MFCD00134037