Cesium standard solution, 1000 mg/L Cs in 5% nitric acid (from Cs₂CO₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13825.AD 13825.AE 13825.AP
13825.ADEA 68.6 EUR
13825.AD 13825.AE 13825.AP
Cesium standard solution, 1000 mg/L Cs in 5% nitric acid (from Cs₂CO₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Cesium standaarden
Molecular Formula: Cs₂CO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134037
Certificaten: Traceable to NIST