Cesium standard solution, 1000 mg/L Cs in 5% nitric acid (from Cs₂CO₃), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88058.AE 88058.AP
88058.AEEA 47.5 EUR
88058.AE 88058.AP
Cesium standard solution, 1000 mg/L Cs in 5% nitric acid (from Cs₂CO₃), standard for AAS, Specpure®
Standaarden Kationische standaarden Cesium standaarden
Molecular Formula: Cs₂CO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134037
Certificaten: Traceable to NIST