Dysprosium standard solution, 10000 mg/L Dy in 5% nitric acid (from Dy₂O₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14426.AD
14426.AE
14426.AP
14426.ADEA
102
EUR
14426.AD
14426.AE
14426.AP
Dysprosium standard solution, 10000 mg/L Dy in 5% nitric acid (from Dy₂O₃), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Dysprosium standaarden
Molecular Formula: Dy₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010982
Certificaten: Traceable to NIST