Gadolinium standard solution, 1000 mg/L Gd in 5% nitric acid (de Gd?O?), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88065.AE 88065.AP
88065.AEEA 44.8 EUR
88065.AE 88065.AP
Gadolinium standard solution, 1000 mg/L Gd in 5% nitric acid (de Gd?O?), standard for AAS, Specpure®
Standaarden Kationische standaarden Gadolinium standaarden
Molecular Formula: Gd₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011022
Certificaten: Traceable to NIST