Manganese standard solution, 10000 mg/L Mn in 5% nitric acid (from Mn), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14413.AD
14413.AE
14413.AP
14413.ADEA
87.4
EUR
14413.AD
14413.AE
14413.AP
Manganese standard solution, 10000 mg/L Mn in 5% nitric acid (from Mn), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Mangaan standaarden
Molecular Formula: Mn in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011111
Certificaten: Traceable to NIST