Manganese standard solution, 1000 µg/ml Mn in 5% nitric acid (from Mn), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13826.AD
13826.AE
13826.AP
13826.ADEA
55.5
EUR
13826.AD
13826.AE
13826.AP
Manganese standard solution, 1000 µg/ml Mn in 5% nitric acid (from Mn), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Mangaan standaarden
Molecular Formula: Mn in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011111
Certificaten: Traceable to NIST