Chromium standard solution, 1000 mg/L Cr in 5% nitric acid (from Cr(NO₃)₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
38728.AD 38728.AE 38728.AP
38728.ADEA 72.8 EUR
38728.AD 38728.AE 38728.AP
Chromium standard solution, 1000 mg/L Cr in 5% nitric acid (from Cr(NO₃)₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Chroom standaarden
Molecular Formula: Cr(NO₃)₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00010944
Certificaten: Traceable to NIST