Magnesium standard solution, 1000 mg/L Mg in 5% nitric acid (from Mg), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88077.AE 88077.AP
88077.AEEA 39.5 EUR
88077.AE 88077.AP
Magnesium standard solution, 1000 mg/L Mg in 5% nitric acid (from Mg), standard for AAS, Specpure®
Standaarden Kationische standaarden Magnesium standaarden
Molecular Formula: Mg in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085308
Certificaten: Traceable to NIST