Silver standard solution, 10000 mg/L Ag in 5% nitric acid (from Ag), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14418.AE
14418.AD
14418.AP
14418.AEEA
164
EUR
14418.AE
14418.AD
14418.AP
Silver standard solution, 10000 mg/L Ag in 5% nitric acid (from Ag), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Zilver standaarden
Molecular Formula: Ag in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00003397
Certificaten: Traceable to NIST