Cerium standard solution, 10000 mg/L Ce in 5% nitric acid (from Ce), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14371.AD 14371.AE 14371.AP
14371.ADEA 83.8 EUR
14371.AD 14371.AE 14371.AP
Cerium standard solution, 10000 mg/L Ce in 5% nitric acid (from Ce), standard for ICP, Specpure®
Standaarden Kationische standaarden Cerium standaarden
Molecular Formula: Ce in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010924
Certificaten: Traceable to NIST
Order Now
Specificaties
Environmentally Preferable
MDL No. MFCD00010924