Cerium standard solution, 1000 mg/L Ce in 5% nitric acid (from Ce), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13815.AD 13815.AE
13815.ADEA 51.1 EUR
13815.AD 13815.AE
Cerium standard solution, 1000 mg/L Ce in 5% nitric acid (from Ce), standard for ICP, Specpure®
Standaarden Kationische standaarden Cerium standaarden
Molecular Formula: Ce in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010924
Certificaten: Traceable to NIST