Cobalt standard solution, 10000 mg/L Co in 5% nitric acid (from Co), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14390.AD 14390.AE 14390.AP
14390.ADEA 92.4 EUR
14390.AD 14390.AE 14390.AP
Cobalt standard solution, 10000 mg/L Co in 5% nitric acid (from Co), standard for ICP, Specpure®
Standaarden Kationische standaarden Kobalt standaarden
Molecular Formula: Co in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010935
Certificaten: Traceable to NIST