Cobalt standard solution, 1000 mg/L Co in 5% nitric acid (from Co), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13828.AD
13828.AE
13828.AP
13828.ADEA
51.5
EUR
13828.AD
13828.AE
13828.AP
Cobalt standard solution, 1000 mg/L Co in 5% nitric acid (from Co), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Kobalt standaarden
Molecular Formula: Co in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010935
Certificaten: Traceable to NIST