Gallium standard solution, 10000 mg/L Ga in 5% nitric acid with hydrochloric acid (max. 1%) (from Ga), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14427.AD
14427.AE
14427.AP
14427.ADEA
103
EUR
14427.AD
14427.AE
14427.AP
Gallium standard solution, 10000 mg/L Ga in 5% nitric acid with hydrochloric acid (max. 1%) (from Ga), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Gallium standaarden
Molecular Formula: Ga in 5% HNO₃/tr. HCl
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134045
Certificaten: Traceable to NIST