Gold standard solution, 10000 mg/L Au in 20% hydrochloric acid (from Au), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14392.AD 14392.AE 14392.AP
14392.ADEA 382 EUR
14392.AD 14392.AE 14392.AP
Gold standard solution, 10000 mg/L Au in 20% hydrochloric acid (from Au), standard for ICP, Specpure®
Standaarden Kationische standaarden Goud standaarden
Molecular Formula: Au in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00003436
Certificaten: Traceable to NIST