Indium standard solution, 10000 mg/L In in 5% nitric acid (from In), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14411.AD 14411.AE 14411.AP
14411.ADEA 132 EUR
14411.AD 14411.AE 14411.AP
Indium standard solution, 10000 mg/L In in 5% nitric acid (from In), standard for ICP, Specpure®
Standaarden Kationische standaarden Indium standaarden
Molecular Formula: In in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134048
Traceable to NIST
Certificaten: Traceable to NIST