Indium standard solution, 1000 mg/L In in 5% nitric acid (from In), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13846.AD
13846.AE
13846.AP
13846.ADEA
73.5
EUR
13846.AD
13846.AE
13846.AP
Indium standard solution, 1000 mg/L In in 5% nitric acid (from In), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Indium standaarden
Molecular Formula: In in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134048
Certificaten: Traceable to NIST