Indium standard solution, 1000 mg/L In in 5% nitric acid (from In), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88071.AE 88071.AP
88071.AEEA 49.8 EUR
88071.AE 88071.AP
Indium standard solution, 1000 mg/L In in 5% nitric acid (from In), standard for AAS, Specpure®
Standaarden Kationische standaarden Indium standaarden
Molecular Formula: In in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134048
Certificaten: Traceable to NIST