Lithium standard solution, 1000 mg/L Li in 5% nitric acid (from Li₂CO₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13821.AD
13821.AE
13821.AP
13821.ADEA
53.5
EUR
13821.AD
13821.AE
13821.AP
Lithium standard solution, 1000 mg/L Li in 5% nitric acid (from Li₂CO₃), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Lithium standaarden
Molecular Formula: Li₂CO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134051
Certificaten: Traceable to NIST