Lithium standard solution, 1000 mg/L Li in 5% nitric acid (from Li₂CO₃), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88076.AE 88076.AP
88076.AEEA 38.6 EUR
88076.AE 88076.AP
Lithium standard solution, 1000 mg/L Li in 5% nitric acid (from Li₂CO₃), standard for AAS, Specpure®
Standaarden Kationische standaarden Lithium standaarden
Molecular Formula: Li₂CO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134051
Certificaten: Traceable to NIST