Molybdenum standard solution, 1000 mg/L Mo in ammonium hydroxide solution (max. 1%) (from (NH₄)₂MoO₄), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
38719.AD
38719.AE
38719.ADEA
68.1
EUR
38719.AD
38719.AE
Molybdenum standard solution, 1000 mg/L Mo in ammonium hydroxide solution (max. 1%) (from (NH₄)₂MoO₄), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Molybdeen standaarden
Molecular Formula: (NH₄)₂MoO₄ in 1% NH₄OH
Physical Form: Liquid
Odour: Ammonia-like
MDL No.: MFCD00003465
Certificaten: Traceable to NIST