Sodium standard solution, 10000 mg/L Na in 5% nitric acid (from Na₂CO₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14400.ADEA 94.6 EUR
14400.AD 14400.AE
Sodium standard solution, 10000 mg/L Na in 5% nitric acid (from Na₂CO₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Natrium standaarden
Molecular Formula: Na₂CO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085307
Certificaten: Traceable to NIST