Sodium standard solution, 1000 mg/L Na in 5% nitric acid (from Na₂CO₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13832.AD
13832.AE
13832.AP
13832.ADEA
53.5
EUR
13832.AD
13832.AE
13832.AP
Sodium standard solution, 1000 mg/L Na in 5% nitric acid (from Na₂CO₃), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Natrium standaarden
Molecular Formula: Na₂CO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085307
Certificaten: Traceable to NIST