Sodium standard solution, 1000 mg/L Na in 5% nitric acid (from Na₂CO₃), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88097.AE 88097.AP
88097.AEEA 41.4 EUR
88097.AE 88097.AP
Sodium standard solution, 1000 mg/L Na in 5% nitric acid (from Na₂CO₃), standard for AAS, Specpure®
Standaarden Kationische standaarden Natrium standaarden
Molecular Formula: Na₂CO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085307
Certificaten: Traceable to NIST