Nickel standard solution, 1000 mg/L Ni in 5% nitric acid (from Ni), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13839.AD 13839.AE 13839.AP
13839.ADEA 54.4 EUR
13839.AD 13839.AE 13839.AP
Nickel standard solution, 1000 mg/L Ni in 5% nitric acid (from Ni), standard for ICP, Specpure®
Standaarden Kationische standaarden Nikkel standaarden
Molecular Formula: Ni in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011137
Certificaten: Traceable to NIST