Palladium standard solution, 1000 mg/L Pd in 20% hydrochloric acid (from Pd), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88085.AE 88085.AP
88085.AEEA 87.6 EUR
88085.AE 88085.AP
Palladium standard solution, 1000 mg/L Pd in 20% hydrochloric acid (from Pd), standard for AAS, Specpure®
Standaarden Kationische standaarden Palladium standaarden
Molecular Formula: Pd in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD03613603
Certificaten: Traceable to NIST