Mercury standard solution, 10000 mg/L Hg in 5% nitric acid (from Hg), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14395.AD
14395.AE
14395.AP
14395.ADEA
86.9
EUR
14395.AD
14395.AE
14395.AP
Mercury standard solution, 10000 mg/L Hg in 5% nitric acid (from Hg), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Kwik standaarden
Molecular Formula: Hg in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011035
Certificaten: Traceable to NIST