Mercury standard solution, 1000 mg/L Hg in 5% nitric acid (from Hg), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13865.AEEA 75.1 EUR
13865.AE 13865.AD
Mercury standard solution, 1000 mg/L Hg in 5% nitric acid (from Hg), standard for ICP, Specpure®
Standaarden Kationische standaarden Kwik standaarden
Molecular Formula: Hg in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011035
Certificaten: Traceable to NIST
Order Now
Specificaties
Environmentally Preferable
MDL No. MFCD00011035