Ruthenium standard solution, 1000 mg/L Ru in 20% hydrochloric acid (from RuCl₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
35767.AD
35767.AE
35767.AP
35767.ADEA
102
EUR
35767.AD
35767.AE
35767.AP
Ruthenium standard solution, 1000 mg/L Ru in 20% hydrochloric acid (from RuCl₃), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Ruthenium standaarden
Molecular Formula: RuCl₃ in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011207
Certificaten: Traceable to NIST