Samarium standard solution, 10000 mg/L Sm in 5% nitric acid (from Sm₂O₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14417.AE 14417.AD 14417.AP
14417.AEEA 144 EUR
14417.AE 14417.AD 14417.AP
Samarium standard solution, 10000 mg/L Sm in 5% nitric acid (from Sm₂O₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Samarium standaarden
Molecular Formula: Sm₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011233
Certificaten: Traceable to NIST