Samarium standard solution, 1000 mg/L Sm in 5% nitric acid (from Sm₂O₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13854.ADEA 70.2 EUR
13854.AD 13854.AE
Samarium standard solution, 1000 mg/L Sm in 5% nitric acid (from Sm₂O₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Samarium standaarden
Molecular Formula: Sm₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011233
Certificaten: Traceable to NIST