Strontium standard solution, 10000 mg/L Sr in 5% nitric acid (from Sr(NO₃)₂), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14382.AD 14382.AE 14382.AP
14382.ADEA 105 EUR
14382.AD 14382.AE 14382.AP
Strontium standard solution, 10000 mg/L Sr in 5% nitric acid (from Sr(NO₃)₂), standard for ICP, Specpure®
Standaarden Kationische standaarden Strontium standaarden
Molecular Formula: Sr(NO₃)₂ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134060
Certificaten: Traceable to NIST