Strontium standard solution, 1000 mg/L Sr in 5% nitric acid (from Sr(NO₃)₂), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88098.AE 88098.AP
88098.AEEA 41.9 EUR
88098.AE 88098.AP
Strontium standard solution, 1000 mg/L Sr in 5% nitric acid (from Sr(NO₃)₂), standard for AAS, Specpure®
Standaarden Kationische standaarden Strontium standaarden
Molecular Formula: Sr(NO₃)₂ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134060
Certificaten: Traceable to NIST