Vanadium standard solution, 10000 mg/L V in 5% nitric acid (from V₂O₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14385.AD
14385.AE
14385.AP
14385.ADEA
102
EUR
14385.AD
14385.AE
14385.AP
Vanadium standard solution, 10000 mg/L V in 5% nitric acid (from V₂O₃), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Vanadium standaarden
Molecular Formula: V₂O₅ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011453
Certificaten: Traceable to NIST