Vanadium standard solution, 1000 mg/L V in 5% nitric acid (from V₂O₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13850.AD 13850.AE 13850.AP
13850.ADEA 52.9 EUR
13850.AD 13850.AE 13850.AP
Vanadium standard solution, 1000 mg/L V in 5% nitric acid (from V₂O₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Vanadium standaarden
Molecular Formula: V₂O₅ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011453
Certificaten: Traceable to NIST
Order Now
Specificaties
Environmentally Preferable
MDL No. MFCD00011453