Vanadium standard solution, 1000 mg/L V in 5% nitric acid (from V₂O₃), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88116.AE 88116.AP
88116.AEEA 51.1 EUR
88116.AE 88116.AP
Vanadium standard solution, 1000 mg/L V in 5% nitric acid (from V₂O₃), standard for AAS, Specpure®
Standaarden Kationische standaarden Vanadium standaarden
Molecular Formula: V₂O₅ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011453
Certificaten: Traceable to NIST