Zinc standard solution, 1000 mg/L Zn in 5% nitric acid (from Zn), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13835.AD 13835.AE 13835.AP
13835.ADEA 52.9 EUR
13835.AD 13835.AE 13835.AP
Zinc standard solution, 1000 mg/L Zn in 5% nitric acid (from Zn), standard for ICP, Specpure®
Standaarden Kationische standaarden Zink standaarden
Molecular Formula: Zn in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011291
Certificaten: Traceable to NIST